Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGAGAAAGCAGGGTGGGTATTA[G/T]AAGGGGAGAAGGGCCCCCACCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605725 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRX PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PRX - periaxin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020956.2 | Intron | NP_066007.1 | ||||
NM_181882.2 | Intron | NP_870998.2 | ||||
XM_011527171.2 | Intron | XP_011525473.1 | ||||
XM_017027046.1 | Intron | XP_016882535.1 | ||||
XM_017027047.1 | Intron | XP_016882536.1 |