Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCCCACCGACACCAGCGAGAGTG[C/G]AGTCGTAACCGTCACGTGACCGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615698 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf47 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C19orf47 - chromosome 19 open reading frame 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256440.1 | 153 | Intron | NP_001243369.1 | |||
NM_001256441.1 | 153 | Intron | NP_001243370.1 | |||
XM_005258520.2 | 153 | Intron | XP_005258577.1 | |||
XM_011526460.2 | 153 | Intron | XP_011524762.1 | |||
XM_017026291.1 | 153 | Intron | XP_016881780.1 | |||
XM_017026292.1 | 153 | Intron | XP_016881781.1 | |||
XM_017026293.1 | 153 | Intron | XP_016881782.1 | |||
XM_017026294.1 | 153 | Intron | XP_016881783.1 | |||
XM_017026295.1 | 153 | Intron | XP_016881784.1 |
PLD3 - phospholipase D family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031696.3 | 153 | UTR 5 | NP_001026866.1 | |||
NM_001291311.1 | 153 | Intron | NP_001278240.1 | |||
NM_012268.3 | 153 | UTR 5 | NP_036400.2 | |||
XM_005258704.1 | 153 | Intron | XP_005258761.1 | |||
XM_005258707.4 | 153 | UTR 5 | XP_005258764.1 | |||
XM_005258708.3 | 153 | Intron | XP_005258765.1 | |||
XM_005258709.4 | 153 | Intron | XP_005258766.1 | |||
XM_005258710.4 | 153 | UTR 5 | XP_005258767.1 | |||
XM_006723122.1 | 153 | UTR 5 | XP_006723185.1 | |||
XM_011526692.1 | 153 | UTR 5 | XP_011524994.1 | |||
XM_011526693.1 | 153 | Intron | XP_011524995.1 | |||
XM_017026546.1 | 153 | UTR 5 | XP_016882035.1 | |||
XM_017026547.1 | 153 | Intron | XP_016882036.1 | |||
XM_017026548.1 | 153 | UTR 5 | XP_016882037.1 | |||
XM_017026549.1 | 153 | UTR 5 | XP_016882038.1 |
Set Membership: |
Validated |