Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611840 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ABHD8 - abhydrolase domain containing 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DDA1 - DET1 and DDB1 associated 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL34 - mitochondrial ribosomal protein L34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023937.3 | 568 | UTR 3 | NP_076426.1 | |||
XM_011528189.2 | 568 | UTR 3 | XP_011526491.1 |
Set Membership: |
HapMap Validated |