Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAATGGTTGTTATATTCTGCCAGA[C/T]AGACTCTTTCTAAAAACACAATTAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605792 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEPT4-AS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SEPT4-AS1 - SEPT4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX14 - testis expressed 14, intercellular bridge forming factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201457.1 | Intron | NP_001188386.1 | ||||
NM_031272.4 | Intron | NP_112562.3 | ||||
NM_198393.3 | Intron | NP_938207.2 | ||||
XM_011525029.2 | Intron | XP_011523331.1 | ||||
XM_017024860.1 | Intron | XP_016880349.1 | ||||
XM_017024861.1 | Intron | XP_016880350.1 |