Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCAGGACCTAGTATACAGGATGC[A/C]CTCAGGAAATATGTGCTGTACAGAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608300 MIM: 600781 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAGS PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
NAGS - N-acetylglutamate synthase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PYY - peptide YY | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004160.5 | Intron | NP_004151.3 | ||||
XM_011525035.1 | Intron | XP_011523337.1 |
TMEM101 - transmembrane protein 101 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304813.1 | Intron | NP_001291742.1 | ||||
NM_001304814.1 | Intron | NP_001291743.1 | ||||
NM_032376.3 | Intron | NP_115752.1 | ||||
XM_005257737.2 | Intron | XP_005257794.1 | ||||
XM_011525346.1 | Intron | XP_011523648.1 | ||||
XM_011525347.1 | Intron | XP_011523649.1 | ||||
XM_011525349.1 | Intron | XP_011523651.1 | ||||
XM_011525351.1 | Intron | XP_011523653.1 | ||||
XM_011525353.1 | Intron | XP_011523655.1 |
Set Membership: |
HapMap |