Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602796 MIM: 602808 MIM: 602817 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIST1H2AM PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HIST1H2AM - histone cluster 1, H2am | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H2BO - histone cluster 1, H2bo | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H3J - histone cluster 1, H3j | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003535.2 | Intron | NP_003526.1 |
Set Membership: |
HapMap |