Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGCCGACAGCACCTGGCAGTG[C/T]GGATAAGGGCATGGCCTGCCCTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612482 MIM: 603555 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF43 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RNF43 - ring finger protein 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305544.1 | 3028 | Silent Mutation | NP_001292473.1 | |||
NM_001305545.1 | 3028 | Silent Mutation | NP_001292474.1 | |||
NM_017763.5 | 3028 | Silent Mutation | NP_060233.3 | |||
XM_011524955.2 | 3028 | Silent Mutation | XP_011523257.1 | |||
XM_011524956.2 | 3028 | Silent Mutation | XP_011523258.1 | |||
XM_017024800.1 | 3028 | Silent Mutation | XP_016880289.1 |
SUPT4H1 - SPT4 homolog, DSIF elongation factor subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPOAP1-AS1 - TSPOAP1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |