Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604264 MIM: 610068 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CELSR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CELSR3 - cadherin EGF LAG seven-pass G-type receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6824 - microRNA 6824 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC26A6 - solute carrier family 26 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040454.1 | Intron | NP_001035544.1 | ||||
NM_001281732.1 | Intron | NP_001268661.1 | ||||
NM_001281733.1 | Intron | NP_001268662.1 | ||||
NM_022911.2 | Intron | NP_075062.2 | ||||
NM_134263.2 | Intron | NP_599025.2 | ||||
NM_134426.2 | Intron | NP_602298.2 |
TMEM89 - transmembrane protein 89 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |