Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605015 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF214 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZNF214 - zinc finger protein 214 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013249.2 | 2838 | Intron | NP_037381.2 | |||
XM_005253128.3 | 2838 | UTR 3 | XP_005253185.1 | |||
XM_005253129.3 | 2838 | UTR 3 | XP_005253186.1 | |||
XM_006718308.3 | 2838 | UTR 3 | XP_006718371.1 | |||
XM_006718309.3 | 2838 | UTR 3 | XP_006718372.1 | |||
XM_011520355.2 | 2838 | UTR 3 | XP_011518657.1 | |||
XM_011520357.2 | 2838 | UTR 3 | XP_011518659.1 | |||
XM_017018269.1 | 2838 | UTR 3 | XP_016873758.1 |