Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAGGGAGGCAGCCTGCAGCCTC[A/G]TCTGTCACCAGGTCCCCTTGGAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605759 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ASB2 - ankyrin repeat and SOCS box containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202429.1 | 2284 | UTR 3 | NP_001189358.1 | |||
NM_016150.4 | 2284 | UTR 3 | NP_057234.2 | |||
XM_005267758.3 | 2284 | UTR 3 | XP_005267815.1 | |||
XM_011536834.2 | 2284 | UTR 3 | XP_011535136.1 | |||
XM_011536835.2 | 2284 | UTR 3 | XP_011535137.1 | |||
XM_017021369.1 | 2284 | UTR 3 | XP_016876858.1 |
FAM181A - family with sequence similarity 181 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM181A-AS1 - FAM181A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |