Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116930 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCAM1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NCAM1 - neural cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000615.6 | 3610 | Intron | NP_000606.3 | |||
NM_001076682.3 | 3610 | UTR 3 | NP_001070150.1 | |||
NM_001242607.1 | 3610 | Intron | NP_001229536.1 | |||
NM_001242608.1 | 3610 | UTR 3 | NP_001229537.1 | |||
NM_181351.4 | 3610 | Intron | NP_851996.2 |
NCAM1-AS1 - NCAM1 antisense RNA1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |