Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGATGATGCTGATCTTGAACATC[A/T]TCTGACACACAGTAGGTACCCTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607425 MIM: 180240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GJD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GJD3 - gap junction protein delta 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152219.3 | 3937 | UTR 3 | NP_689343.3 |
RARA - retinoic acid receptor alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |