Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612074 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRRT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRRT4 - proline rich transmembrane protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114726.2 | 1640 | UTR 3 | NP_001108198.2 | |||
NM_001174164.1 | 1640 | Missense Mutation | CCG,CTG | P,L 826 | NP_001167635.1 | |
XM_005250342.3 | 1640 | Missense Mutation | CCG,CTG | P,L 826 | XP_005250399.1 | |
XM_005250343.3 | 1640 | Missense Mutation | CCG,CTG | P,L 826 | XP_005250400.1 | |
XM_006715985.3 | 1640 | Missense Mutation | CCG,CTG | P,L 826 | XP_006716048.1 | |
XM_017012205.1 | 1640 | Missense Mutation | CCG,CTG | P,L 826 | XP_016867694.1 | |
XM_017012206.1 | 1640 | UTR 3 | XP_016867695.1 |
RBM28 - RNA binding motif protein 28 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |