Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600289 MIM: 608480 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAPK14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MAPK14 - mitogen-activated protein kinase 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001315.2 | Intron | NP_001306.1 | ||||
NM_139012.2 | Intron | NP_620581.1 | ||||
NM_139013.2 | Intron | NP_620582.1 | ||||
NM_139014.2 | Intron | NP_620583.1 | ||||
XM_006714998.3 | Intron | XP_006715061.1 | ||||
XM_011514310.2 | Intron | XP_011512612.2 | ||||
XM_017010299.1 | Intron | XP_016865788.1 | ||||
XM_017010300.1 | Intron | XP_016865789.1 | ||||
XM_017010301.1 | Intron | XP_016865790.1 | ||||
XM_017010302.1 | Intron | XP_016865791.1 | ||||
XM_017010303.1 | Intron | XP_016865792.1 | ||||
XM_017010304.1 | Intron | XP_016865793.1 |
SLC26A8 - solute carrier family 26 member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |