Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608760 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
VGLL4 - vestigial like family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128219.2 | Intron | NP_001121691.1 | ||||
NM_001128220.2 | Intron | NP_001121692.1 | ||||
NM_001128221.2 | Intron | NP_001121693.1 | ||||
NM_001284390.1 | Intron | NP_001271319.1 | ||||
NM_001284391.1 | Intron | NP_001271320.1 | ||||
NM_014667.3 | Intron | NP_055482.2 | ||||
XM_011534267.2 | Intron | XP_011532569.1 | ||||
XM_011534268.1 | Intron | XP_011532570.1 | ||||
XM_011534269.1 | Intron | XP_011532571.1 | ||||
XM_017007541.1 | Intron | XP_016863030.1 | ||||
XM_017007542.1 | Intron | XP_016863031.1 |
Set Membership: |
HapMap |