Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611445 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNM3 - dynamin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136127.2 | Intron | NP_001129599.1 | ||||
NM_001278252.1 | Intron | NP_001265181.1 | ||||
NM_015569.4 | Intron | NP_056384.2 | ||||
XM_005245079.1 | Intron | XP_005245136.1 | ||||
XM_005245080.1 | Intron | XP_005245137.1 | ||||
XM_006711268.2 | Intron | XP_006711331.1 | ||||
XM_017000976.1 | Intron | XP_016856465.1 | ||||
XM_017000977.1 | Intron | XP_016856466.1 | ||||
XM_017000978.1 | Intron | XP_016856467.1 | ||||
XM_017000979.1 | Intron | XP_016856468.1 | ||||
XM_017000980.1 | Intron | XP_016856469.1 | ||||
XM_017000981.1 | Intron | XP_016856470.1 | ||||
XM_017000982.1 | Intron | XP_016856471.1 | ||||
XM_017000983.1 | Intron | XP_016856472.1 | ||||
XM_017000984.1 | Intron | XP_016856473.1 | ||||
XM_017000985.1 | Intron | XP_016856474.1 | ||||
XM_017000986.1 | Intron | XP_016856475.1 | ||||
XM_017000987.1 | Intron | XP_016856476.1 | ||||
XM_017000988.1 | Intron | XP_016856477.1 | ||||
XM_017000989.1 | Intron | XP_016856478.1 | ||||
XM_017000990.1 | Intron | XP_016856479.1 | ||||
XM_017000991.1 | Intron | XP_016856480.1 | ||||
XM_017000992.1 | Intron | XP_016856481.1 |