Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGGGTTTTTCCGGCCACTCAGTT[C/A]TGCCACCGTCACTGAGAAGCTCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
POLA2 - DNA polymerase alpha 2, accessory subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002689.3 | 54 | UTR 5 | NP_002680.2 | |||
XM_011544877.2 | 54 | UTR 5 | XP_011543179.1 | |||
XM_011544878.2 | 54 | UTR 5 | XP_011543180.1 | |||
XM_011544879.1 | 54 | UTR 5 | XP_011543181.1 | |||
XM_011544881.2 | 54 | UTR 5 | XP_011543183.1 | |||
XM_017017452.1 | 54 | UTR 5 | XP_016872941.1 |
Set Membership: |
Validated |