Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTGTCCTGGTACTCCAAAATGAT[A/T]CAGAGCCCCAGCAACAAATATCAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609767 | ||||||||||||||||||||
Literature Links: |
SLC25A28 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC25A28 - solute carrier family 25 member 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031212.3 | Intron | NP_112489.3 | ||||
XM_005270208.1 | Intron | XP_005270265.1 | ||||
XM_005270211.4 | Intron | XP_005270268.1 | ||||
XM_006718005.3 | Intron | XP_006718068.1 | ||||
XM_006718006.3 | Intron | XP_006718069.1 | ||||
XM_011540239.2 | Intron | XP_011538541.1 | ||||
XM_011540240.1 | Intron | XP_011538542.1 | ||||
XM_011540244.2 | Intron | XP_011538546.1 | ||||
XM_017016741.1 | Intron | XP_016872230.1 | ||||
XM_017016742.1 | Intron | XP_016872231.1 |