Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602509 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOLGA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GOLGA4 - golgin A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172713.1 | Intron | NP_001166184.1 | ||||
NM_002078.4 | Intron | NP_002069.2 | ||||
XM_005265069.3 | Intron | XP_005265126.1 | ||||
XM_005265070.3 | Intron | XP_005265127.1 | ||||
XM_005265071.3 | Intron | XP_005265128.1 | ||||
XM_005265072.3 | Intron | XP_005265129.1 | ||||
XM_005265073.1 | Intron | XP_005265130.1 | ||||
XM_005265074.1 | Intron | XP_005265131.1 | ||||
XM_005265075.1 | Intron | XP_005265132.1 | ||||
XM_006713110.2 | Intron | XP_006713173.1 | ||||
XM_011533598.2 | Intron | XP_011531900.1 | ||||
XM_017006184.1 | Intron | XP_016861673.1 | ||||
XM_017006185.1 | Intron | XP_016861674.1 | ||||
XM_017006186.1 | Intron | XP_016861675.1 | ||||
XM_017006187.1 | Intron | XP_016861676.1 | ||||
XM_017006188.1 | Intron | XP_016861677.1 |
LOC152048 - uncharacterized FLJ31715 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |