Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTTTCTGGAACACAATGGCCAAA[A/G]TAGAAGACATCTGGTTTATATGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603296 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DNAJB14 - DnaJ heat shock protein family (Hsp40) member B14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031723.3 | Intron | NP_001026893.1 | ||||
NM_001278310.1 | Intron | NP_001265239.1 | ||||
NM_001278311.1 | Intron | NP_001265240.1 | ||||
XM_011532262.2 | Intron | XP_011530564.1 | ||||
XM_011532263.2 | Intron | XP_011530565.1 | ||||
XM_011532265.2 | Intron | XP_011530567.1 | ||||
XM_017008628.1 | Intron | XP_016864117.1 |
LAMTOR3 - late endosomal/lysosomal adaptor, MAPK and MTOR activator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |