Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGCTTTCAGACTGATCCCCCCT[A/C]ATATGCCTCTGTCCTCACAGGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606360 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WNT8A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WNT8A - Wnt family member 8A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300938.1 | Intron | NP_001287867.1 | ||||
NM_001300939.1 | Intron | NP_001287868.1 | ||||
NM_058244.3 | Intron | NP_490645.1 | ||||
XM_011543625.2 | Intron | XP_011541927.2 | ||||
XM_017009824.1 | Intron | XP_016865313.1 | ||||
XM_017009825.1 | Intron | XP_016865314.1 | ||||
XM_017009826.1 | Intron | XP_016865315.1 |
Set Membership: |
HapMap |