Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATAAAAATGGTTCCATTTTATCA[A/G]GTAGGTTTTTCTTTTAAAGGGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610686 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBXN2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
UBXN2B - UBX domain protein 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077619.1 | Intron | NP_001071087.1 | ||||
XM_005251165.2 | Intron | XP_005251222.1 | ||||
XM_005251166.1 | Intron | XP_005251223.1 |
Set Membership: |
Validated |