Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603475 MIM: 606023 MIM: 600044 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CHRD - chordin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR2H - RNA polymerase II subunit H | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THPO - thrombopoietin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000460.3 | 1376 | UTR 3 | NP_000451.1 | |||
NM_001177597.2 | 1376 | UTR 3 | NP_001171068.1 | |||
NM_001177598.2 | 1376 | UTR 3 | NP_001171069.1 | |||
NM_001289997.1 | 1376 | UTR 3 | NP_001276926.1 | |||
NM_001289998.1 | 1376 | UTR 3 | NP_001276927.1 | |||
NM_001290003.1 | 1376 | UTR 3 | NP_001276932.1 | |||
NM_001290022.1 | 1376 | UTR 3 | NP_001276951.1 | |||
NM_001290026.1 | 1376 | UTR 3 | NP_001276955.1 | |||
NM_001290027.1 | 1376 | UTR 3 | NP_001276956.1 | |||
NM_001290028.1 | 1376 | UTR 3 | NP_001276957.1 | |||
XM_017007107.1 | 1376 | UTR 3 | XP_016862596.1 |