Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCCTCCTTCATGTATTCCCTCT[C/G]TACGGAGAGTAGGCAGAGAGATGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
44 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603492 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLAMF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLAMF1 - signaling lymphocytic activation molecule family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003037.3 | Intron | NP_003028.1 | ||||
XM_005245456.3 | Intron | XP_005245513.1 | ||||
XM_011509905.2 | Intron | XP_011508207.1 | ||||
XM_017002129.1 | Intron | XP_016857618.1 | ||||
XM_017002130.1 | Intron | XP_016857619.1 | ||||
XM_017002131.1 | Intron | XP_016857620.1 |
Set Membership: |
HapMap |