Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGCGGCAGTGGCCTCGCAGGGCG[C/T]TGGGTCCCTCTCCCCAGCTCTCCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610800 | ||||||||||||||||||||
Literature Links: |
ATG10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATG10 - autophagy related 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131028.1 | 196 | UTR 5 | NP_001124500.1 | |||
NM_031482.4 | 196 | UTR 5 | NP_113670.1 | |||
XM_005248610.4 | 196 | Intron | XP_005248667.1 | |||
XM_005248611.4 | 196 | Intron | XP_005248668.1 | |||
XM_005248612.3 | 196 | UTR 5 | XP_005248669.1 | |||
XM_011543660.2 | 196 | Intron | XP_011541962.1 | |||
XM_011543661.2 | 196 | Intron | XP_011541963.1 | |||
XM_017009944.1 | 196 | UTR 5 | XP_016865433.1 |