Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600951 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TERF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TERF1 - telomeric repeat binding factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003218.3 | Intron | NP_003209.2 | ||||
NM_017489.2 | Intron | NP_059523.2 | ||||
XM_005251291.3 | Intron | XP_005251348.1 | ||||
XM_005251292.3 | Intron | XP_005251349.1 | ||||
XM_011517582.2 | Intron | XP_011515884.1 | ||||
XM_017013792.1 | Intron | XP_016869281.1 |