Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATCAAGTGCAGTGTCCTTTTAG[G/T]CATGAGATCCTATGTGACTGCATGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605515 | |||||||||||||||||||||||
Literature Links: |
FOXP1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FOXP1 - forkhead box P1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012505.1 | Intron | NP_001012523.1 | ||||
NM_001244808.1 | Intron | NP_001231737.1 | ||||
NM_001244810.1 | Intron | NP_001231739.1 | ||||
NM_001244812.1 | Intron | NP_001231741.1 | ||||
NM_001244813.1 | Intron | NP_001231742.1 | ||||
NM_001244814.1 | Intron | NP_001231743.1 | ||||
NM_001244815.1 | Intron | NP_001231744.1 | ||||
NM_001244816.1 | Intron | NP_001231745.1 | ||||
NM_032682.5 | Intron | NP_116071.2 | ||||
XM_005264735.3 | Intron | XP_005264792.1 | ||||
XM_005264736.3 | Intron | XP_005264793.1 | ||||
XM_005264737.4 | Intron | XP_005264794.1 | ||||
XM_005264742.3 | Intron | XP_005264799.1 | ||||
XM_006713102.2 | Intron | XP_006713165.1 | ||||
XM_006713103.2 | Intron | XP_006713166.1 | ||||
XM_006713104.2 | Intron | XP_006713167.1 | ||||
XM_011533584.2 | Intron | XP_011531886.1 | ||||
XM_011533585.2 | Intron | XP_011531887.1 | ||||
XM_011533588.2 | Intron | XP_011531890.1 | ||||
XM_017006165.1 | Intron | XP_016861654.1 | ||||
XM_017006166.1 | Intron | XP_016861655.1 | ||||
XM_017006167.1 | Intron | XP_016861656.1 | ||||
XM_017006168.1 | Intron | XP_016861657.1 |