Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614902 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP33 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ARHGAP33 - Rho GTPase activating protein 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172630.1 | 1690 | UTR 5 | NP_001166101.1 | |||
NM_052948.3 | 1690 | Silent Mutation | GCC,GCT | A,A 50 | NP_443180.2 | |
XM_005258488.1 | 1690 | Silent Mutation | GCC,GCT | A,A 50 | XP_005258545.1 | |
XM_006722999.1 | 1690 | Silent Mutation | GCC,GCT | A,A 50 | XP_006723062.1 | |
XM_006723003.2 | 1690 | Intron | XP_006723066.1 | |||
XM_011526416.1 | 1690 | UTR 5 | XP_011524718.1 | |||
XM_011526417.1 | 1690 | UTR 5 | XP_011524719.1 | |||
XM_011526418.1 | 1690 | UTR 5 | XP_011524720.1 | |||
XM_011526419.1 | 1690 | UTR 5 | XP_011524721.1 | |||
XM_011526420.1 | 1690 | UTR 5 | XP_011524722.1 | |||
XM_011526421.2 | 1690 | Silent Mutation | GCC,GCT | A,A 87 | XP_011524723.1 | |
XM_017026238.1 | 1690 | Silent Mutation | GCC,GCT | A,A 87 | XP_016881727.1 |
PROSER3 - proline and serine rich 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |