Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602352 MIM: 611988 MIM: 176884 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNRH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GNRH2 - gonadotropin releasing hormone 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001310220.1 | 847 | Intron | NP_001297149.1 | |||
NM_001501.1 | 847 | Intron | NP_001492.1 | |||
NM_178331.1 | 847 | Intron | NP_847901.1 | |||
NM_178332.1 | 847 | Intron | NP_847902.1 | |||
XM_017027823.1 | 847 | UTR 5 | XP_016883312.1 |
MRPS26 - mitochondrial ribosomal protein S26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTPRA - protein tyrosine phosphatase, receptor type A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |