Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTTTGTGCATGTATGTGTGGAT[C/G]TATGAGAGGCGTGAGATTGGTATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615875 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928118 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101928118 - uncharacterized LOC101928118 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RWDD3 - RWD domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128142.1 | Intron | NP_001121614.1 | ||||
NM_001199682.1 | Intron | NP_001186611.1 | ||||
NM_001278247.1 | Intron | NP_001265176.1 | ||||
NM_001278248.1 | Intron | NP_001265177.1 | ||||
NM_015485.4 | Intron | NP_056300.2 |
TMEM56-RWDD3 - TMEM56-RWDD3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199691.1 | Intron | NP_001186620.1 |