Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATCATCTTTTTGAAAAGTGGC[A/G]TGAAGTGGTGTGGAACACATCAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
72 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615875 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928118 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
LOC101928118 - uncharacterized LOC101928118 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RWDD3 - RWD domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128142.1 | Intron | NP_001121614.1 | ||||
NM_001199682.1 | Intron | NP_001186611.1 | ||||
NM_001278247.1 | Intron | NP_001265176.1 | ||||
NM_001278248.1 | Intron | NP_001265177.1 | ||||
NM_015485.4 | Intron | NP_056300.2 |
TMEM56-RWDD3 - TMEM56-RWDD3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199691.1 | Intron | NP_001186620.1 |