Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCAGCTCAGGTCCAAGGGCTAG[C/T]GAAACAGACTTTGTGCATTGATGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603826 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NR1H4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NR1H4 - nuclear receptor subfamily 1 group H member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206977.1 | Intron | NP_001193906.1 | ||||
NM_001206978.1 | Intron | NP_001193907.1 | ||||
NM_001206979.1 | Intron | NP_001193908.1 | ||||
NM_001206992.1 | Intron | NP_001193921.1 | ||||
NM_001206993.1 | Intron | NP_001193922.1 | ||||
NM_005123.3 | Intron | NP_005114.1 | ||||
XM_006719719.2 | Intron | XP_006719782.1 | ||||
XM_011539040.2 | Intron | XP_011537342.1 | ||||
XM_011539041.2 | Intron | XP_011537343.1 | ||||
XM_011539042.1 | Intron | XP_011537344.1 |