Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606568 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LZTFL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LZTFL1 - leucine zipper transcription factor like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276378.1 | Intron | NP_001263307.1 | ||||
NM_001276379.1 | Intron | NP_001263308.1 | ||||
NM_020347.3 | Intron | NP_065080.1 | ||||
XM_006713207.2 | Intron | XP_006713270.2 | ||||
XM_011533838.2 | Intron | XP_011532140.1 | ||||
XM_017006644.1 | Intron | XP_016862133.1 | ||||
XM_017006645.1 | Intron | XP_016862134.1 |
Set Membership: |
HapMap Validated |