Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGTGTTCTTAGTTATTGTTCCT[C/G]CTCTGATGGTGCTCCAGGGTCTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600126 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE4A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PDE4A - phosphodiesterase 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111307.1 | Intron | NP_001104777.1 | ||||
NM_001111308.1 | Intron | NP_001104778.1 | ||||
NM_001111309.1 | Intron | NP_001104779.1 | ||||
NM_001243121.1 | Intron | NP_001230050.1 | ||||
NM_006202.2 | Intron | NP_006193.1 | ||||
XM_005259933.2 | Intron | XP_005259990.1 | ||||
XM_011528054.1 | Intron | XP_011526356.1 | ||||
XM_011528055.2 | Intron | XP_011526357.1 | ||||
XM_017026865.1 | Intron | XP_016882354.1 |
Set Membership: |
HapMap Validated |