Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCACATGCCTGGTTCACAGGAGA[A/T]GCTCAATAAACCTGTGGAAAATTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602729 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABRP - gamma-aminobutyric acid type A receptor pi subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291985.1 | Intron | NP_001278914.1 | ||||
NM_014211.2 | Intron | NP_055026.1 | ||||
XM_005265872.2 | Intron | XP_005265929.1 |