Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGAGCTAGAATTTGAACCCAATC[C/T]CAACCCTTTCTGCCTCCGGTAAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607160 MIM: 102565 MIM: 609344 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V1F PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATP6V1F - ATPase H+ transporting V1 subunit F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198909.1 | Intron | NP_001185838.1 | ||||
NM_004231.3 | Intron | NP_004222.2 |
FLNC - filamin C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCP - kielin/chordin-like protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100130705 - uncharacterized LOC100130705 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |