Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCAATTCAATGTTTCAGCTGCTT[G/T]GCTATATCTTAAATTATTTGATGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607567 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJA1P5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAJA1P5 - DnaJ heat shock protein family (Hsp40) member A1 pseudogene 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OLFM3 - olfactomedin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288821.1 | Intron | NP_001275750.1 | ||||
NM_001288823.1 | Intron | NP_001275752.1 | ||||
NM_058170.3 | Intron | NP_477518.2 | ||||
XM_017000240.1 | Intron | XP_016855729.1 |
Set Membership: |
HapMap |