Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAACACTTTAATCACCTTCACGT[A/G]TTTGTGATAACGTTTGTCCTGGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
53 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614827 MIM: 612532 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAJC11 - DnaJ heat shock protein family (Hsp40) member C11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHF13 - PHD finger protein 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THAP3 - THAP domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195752.1 | Intron | NP_001182681.1 | ||||
NM_001195753.1 | Intron | NP_001182682.1 | ||||
NM_138350.3 | Intron | NP_612359.2 | ||||
XM_005263532.3 | Intron | XP_005263589.1 | ||||
XM_011542400.2 | Intron | XP_011540702.1 | ||||
XM_011542401.2 | Intron | XP_011540703.1 | ||||
XM_017002761.1 | Intron | XP_016858250.1 |