Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAGGCCAGTTCGCGTGCTGCCCT[C/G]GGCCGGATCGTCCCCTCAAGCATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616128 MIM: 606044 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EHBP1L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EHBP1L1 - EH domain binding protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099409.1 | Intron | NP_001092879.1 | ||||
XM_005273867.3 | Intron | XP_005273924.1 | ||||
XM_005273868.3 | Intron | XP_005273925.1 | ||||
XM_005273869.3 | Intron | XP_005273926.1 | ||||
XM_005273870.3 | Intron | XP_005273927.1 | ||||
XM_006718486.3 | Intron | XP_006718549.1 | ||||
XM_006718487.3 | Intron | XP_006718550.1 | ||||
XM_011544886.2 | Intron | XP_011543188.1 | ||||
XM_017017467.1 | Intron | XP_016872956.1 | ||||
XM_017017468.1 | Intron | XP_016872957.1 | ||||
XM_017017469.1 | Intron | XP_016872958.1 |
FAM89B - family with sequence similarity 89 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SSSCA1 - Sjogren syndrome/scleroderma autoantigen 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SSSCA1-AS1 - SSSCA1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |