Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCAGTCCTCACTGGGACAACCT[A/G]TGCATTGCTTCCATCCACTCTCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611502 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CENPK - centromere protein K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267038.1 | Intron | NP_001253967.1 | ||||
NM_022145.4 | Intron | NP_071428.2 | ||||
XM_011543534.2 | Intron | XP_011541836.1 | ||||
XM_011543535.2 | Intron | XP_011541837.1 | ||||
XM_011543536.2 | Intron | XP_011541838.1 | ||||
XM_011543537.2 | Intron | XP_011541839.1 | ||||
XM_011543538.2 | Intron | XP_011541840.1 | ||||
XM_011543539.2 | Intron | XP_011541841.1 | ||||
XM_011543540.2 | Intron | XP_011541842.1 | ||||
XM_011543541.2 | Intron | XP_011541843.1 | ||||
XM_011543542.2 | Intron | XP_011541844.1 | ||||
XM_017009690.1 | Intron | XP_016865179.1 | ||||
XM_017009691.1 | Intron | XP_016865180.1 | ||||
XM_017009692.1 | Intron | XP_016865181.1 | ||||
XM_017009693.1 | Intron | XP_016865182.1 | ||||
XM_017009694.1 | Intron | XP_016865183.1 | ||||
XM_017009695.1 | Intron | XP_016865184.1 | ||||
XM_017009696.1 | Intron | XP_016865185.1 | ||||
XM_017009697.1 | Intron | XP_016865186.1 | ||||
XM_017009698.1 | Intron | XP_016865187.1 | ||||
XM_017009699.1 | Intron | XP_016865188.1 | ||||
XM_017009700.1 | Intron | XP_016865189.1 | ||||
XM_017009701.1 | Intron | XP_016865190.1 |