Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACTTGGAAAAGGAAAAGGTAAGG[A/G]AAAATTTGCTTCTCTTTTGTCAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609506 MIM: 604466 MIM: 615258 MIM: 604723 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP27B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYP27B1 - cytochrome P450 family 27 subfamily B member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL1 - methyltransferase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL21B - methyltransferase like 21B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015433.2 | Intron | NP_056248.2 | ||||
NM_206914.1 | Intron | NP_996797.1 |
TSFM - Ts translation elongation factor, mitochondrial | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |