Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGGAAAAGAGGCTTAAAACAAG[A/G]AAACACTGTATTTCCTCCCTCAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606557 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL11A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCL11A - B-cell CLL/lymphoma 11A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018014.3 | Intron | NP_060484.2 | ||||
NM_022893.3 | Intron | NP_075044.2 | ||||
NM_138559.1 | Intron | NP_612569.1 | ||||
XM_011532909.1 | Intron | XP_011531211.1 | ||||
XM_011532910.1 | Intron | XP_011531212.1 | ||||
XM_011532912.1 | Intron | XP_011531214.1 | ||||
XM_017004333.1 | Intron | XP_016859822.1 | ||||
XM_017004334.1 | Intron | XP_016859823.1 | ||||
XM_017004335.1 | Intron | XP_016859824.1 | ||||
XM_017004336.1 | Intron | XP_016859825.1 | ||||
XM_017004337.1 | Intron | XP_016859826.1 | ||||
XM_017004338.1 | Intron | XP_016859827.1 |