Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605140 MIM: 609108 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCT7 - chaperonin containing TCP1 subunit 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXO41 - F-box protein 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080410.2 | 5953 | UTR 3 | NP_001073879.2 | |||
XM_005264158.3 | 5953 | UTR 3 | XP_005264215.1 | |||
XM_006711950.2 | 5953 | UTR 3 | XP_006712013.1 | |||
XM_011532562.1 | 5953 | UTR 3 | XP_011530864.1 | |||
XM_011532563.2 | 5953 | UTR 3 | XP_011530865.1 | |||
XM_017003421.1 | 5953 | UTR 3 | XP_016858910.1 | |||
XM_017003422.1 | 5953 | UTR 3 | XP_016858911.1 | |||
XM_017003423.1 | 5953 | UTR 3 | XP_016858912.1 |