Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164770 MIM: 173410 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSF1R PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CSF1R - colony stimulating factor 1 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDGFRB - platelet derived growth factor receptor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002609.3 | 4201 | Silent Mutation | CCA,CCG | P,P 1084 | NP_002600.1 | |
XM_005268464.2 | 4201 | Silent Mutation | CCA,CCG | P,P 1020 | XP_005268521.1 | |
XM_011537658.1 | 4201 | Silent Mutation | CCA,CCG | P,P 1084 | XP_011535960.1 | |
XM_011537659.1 | 4201 | Silent Mutation | CCA,CCG | P,P 1084 | XP_011535961.1 |
Set Membership: |
HapMap |