Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613529 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP152 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CEP152 - centrosomal protein 152 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001194998.1 | Intron | NP_001181927.1 | ||||
NM_014985.3 | Intron | NP_055800.2 | ||||
XM_006720437.3 | Intron | XP_006720500.1 | ||||
XM_011521373.2 | Intron | XP_011519675.1 | ||||
XM_011521374.2 | Intron | XP_011519676.1 | ||||
XM_011521375.2 | Intron | XP_011519677.1 | ||||
XM_011521378.2 | Intron | XP_011519680.1 | ||||
XM_011521379.2 | Intron | XP_011519681.1 | ||||
XM_011521381.2 | Intron | XP_011519683.1 | ||||
XM_017022014.1 | Intron | XP_016877503.1 | ||||
XM_017022015.1 | Intron | XP_016877504.1 | ||||
XM_017022016.1 | Intron | XP_016877505.1 |
Set Membership: |
HapMap |