Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAGCTCAGCTCTACAGTCCATTA[C/G]CAAGTCTACAAATGCTGCCCTTACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613261 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRTG PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRTG - protogenin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173814.5 | 11848 | UTR 3 | NP_776175.2 | |||
XM_011521459.1 | 11848 | Intron | XP_011519761.1 | |||
XM_011521460.1 | 11848 | Intron | XP_011519762.1 | |||
XM_017022081.1 | 11848 | UTR 3 | XP_016877570.1 | |||
XM_017022082.1 | 11848 | Intron | XP_016877571.1 |