Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAACTGCTCAGCACCAGGACAC[A/G]GAGCTGCATTGCTTGATTCTGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf37 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf37 - chromosome 14 open reading frame 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001872.3 | 10759 | Intron | NP_001001872.2 | |||
NM_001320173.1 | 10759 | Intron | NP_001307102.1 | |||
XM_011536471.2 | 10759 | Intron | XP_011534773.1 | |||
XM_017021029.1 | 10759 | UTR 3 | XP_016876518.1 | |||
XM_017021030.1 | 10759 | UTR 3 | XP_016876519.1 |