Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDSUB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
WDSUB1 - WD repeat, sterile alpha motif and U-box domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128212.2 | Intron | NP_001121684.1 | ||||
NM_001128213.1 | Intron | NP_001121685.1 | ||||
NM_001307994.1 | Intron | NP_001294923.1 | ||||
NM_152528.2 | Intron | NP_689741.2 | ||||
XM_005246355.2 | Intron | XP_005246412.2 | ||||
XM_006712341.3 | Intron | XP_006712404.1 | ||||
XM_011510753.2 | Intron | XP_011509055.1 | ||||
XM_011510754.2 | Intron | XP_011509056.1 | ||||
XM_017003481.1 | Intron | XP_016858970.1 | ||||
XM_017003482.1 | Intron | XP_016858971.1 | ||||
XM_017003483.1 | Intron | XP_016858972.1 |
Set Membership: |
HapMap Validated |