Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300191 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TSPAN6 - tetraspanin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278740.1 | Intron | NP_001265669.1 | ||||
NM_001278741.1 | Intron | NP_001265670.1 | ||||
NM_001278742.1 | Intron | NP_001265671.1 | ||||
NM_001278743.1 | Intron | NP_001265672.1 | ||||
NM_003270.3 | Intron | NP_003261.1 | ||||
XM_011531018.1 | Intron | XP_011529320.1 |