Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604665 MIM: 607273 | ||||||||||||||||||||
Literature Links: |
COPS3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COPS3 - COP9 signalosome subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199125.1 | 1353 | UTR 3 | NP_001186054.1 | |||
NM_001316354.1 | 1353 | UTR 3 | NP_001303283.1 | |||
NM_001316355.1 | 1353 | UTR 3 | NP_001303284.1 | |||
NM_001316356.1 | 1353 | UTR 3 | NP_001303285.1 | |||
NM_001316357.1 | 1353 | UTR 3 | NP_001303286.1 | |||
NM_001316358.1 | 1353 | UTR 3 | NP_001303287.1 | |||
NM_003653.3 | 1353 | UTR 3 | NP_003644.2 | |||
XM_005256837.4 | 1353 | UTR 3 | XP_005256894.1 | |||
XM_005256840.3 | 1353 | UTR 3 | XP_005256897.1 | |||
XM_005256842.4 | 1353 | UTR 3 | XP_005256899.1 | |||
XM_017025246.1 | 1353 | UTR 3 | XP_016880735.1 |
FLCN - folliculin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |